Learn More
Thermo Scientific™ Owl OSP Electrophoresis Power Supplies
Compact, easy-to-use 10-300V and 10-500V power supply units
Description
Description
  • Built-in timer permits unattended runs
  • Four outputs allow four units to be run with clear LED readout
  • Safety features include no-load detection, current/load alarm, and sudden load detection
  • Samples are protected by automatic restart after power failure
  • 300V unit ideal for DNA, RNA, mini SDS-page, pulse field electrophoresis, tank electroblotting, and electroelution
  • 500V unit suited for DNA, RNA, and protein electrophoresis