missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ ADJ1 Adjustable System Accessories and Replacement Parts
Extremely compact at only 16cm wide
Brand: Thermo Scientific™ ADJ1-40G
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- The height of the upper buffer chamber (UBC) may be adjusted by sliding the UBC up or down two support poles allowing gels to vary in height from 14 cm to 40 cm.
- The notched or offset glass plate of the gel sandwich presses against a silicone gasket in the UBC for a leakproof seal.
- Plates are clamped to the UBC with either binder clamps or heavy duty pony clamps.
Specifications
Owl ADJ1 Adjustable Electrophoresis System | |
16 x 14 cm | |
0.18 in. | |
15.7 x 6.2 in. (40 x 16 cm) | |
40 cm | |
6.2 in. | |
16 cm |
Blank Glass Plate | |
Glass | |
0.48 cm | |
15.7 in. | |
6.2 in. | |
16 cm |