missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Orion™ Test Kits for AQUAfast™ IV Colorimeters
Description
- Kits offer premeasured reagents in bar-coded Auto-Test™ cuvettes
- AQUAfast IV Colorimeter has an Auto-ID™ function that reads the bar code to identify the species of interest automatically, then selects proper method, wavelength and time
- Just read results on built-in display
- cuvette is opened only when measurement is taken, so reagent quality is not affected
Specifications
Specifications
| Certifications/Compliance | EPA |
| For Use With (Equipment) | AQUAfast™ IV colorimeter |
| No. of Tests | 30 |
| Range | 0 to 0.4mg/L |
| Test Method | Pyridine-Barbituric Acid |
| Type | AQUAfast™ IV Colorimeter Reagent Kit |
Safety and Handling
Signal Word
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
- P202-Do not handle until all safety precautions have been read and understood.
- P273-Avoid release to the environment.
- P321-Specific treatment (see on this label).
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?