Learn More
Thermo Scientific™ Nunc™ Open Well Upper Structures
Product Code. 11347030
Click to view available options
Quantity:
Case of 100
Unit Size:
Pack of 100
Product Code. 11347030
Brand: Thermo Scientific™ 256665
Only null left
Add to basket
Add to basket
Versatile units in a choice of two materials
Used for bonding to a specialty base material, holding PCR plates, adding agar to each well, or other purposes
  • Nonsterile, black
  • 96- well formats available
  • Polystyrene
  • Nonsterile
  • Without lid
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
No. per Case 100
Plate Compatibility 96-Well Plate
Quantity Case of 100