Learn More
Thermo Scientific™ Nunc™ OmniTray
Product Code. 10164012
Change view
Click to view available options
Culture Area:
84 cm2
Unit Size:
Pack of 90
Product Code. Culture Area unitSize
10164012 84 cm2 Pack of 90
1 options
Product Code. 10164012 Supplier Thermo Scientific™ Supplier No. 140175
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Supports a variety of cell culture applications
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements and colony counting
  • Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Optical clarity makes colonies easy to see
  • Can be frozen at -20°C
Nunc OmniTray acts as:
  • Holder for membrane when dot blotting
  • Container for denaturation, hybridization and washing steps
  • Storage container for membranes
140156 and 140175
  • Feature an enhanced rectangular shape ideal for automatic picking and viewing systems
Dot blotting, denaturation/hybridization/washing, bacterial growth, membrane storage
Compliance
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
TRUSTED_SUSTAINABILITY
Specifications
Culture Area 84 cm2