Learn More
Thermo Scientific™ Nunc™ Caps for TufRol™ Roller Bottles
Vented and non-vented
£366.00 - £610.00
Specifications
No. per Case 500 Cs.
Sterility Sterile
For Use With (Equipment) TufRol roller bottle
View More Specs
Products 2
Product Code Brand Closure Type Price Quantity & Availability  
Product Code Brand Closure Type Price Quantity & Availability  
11997964
View Documents Promotion Details
Thermo Scientific™
112250
Non-vented caps This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11987964
View Documents Promotion Details
Thermo Scientific™
111250
Vented
£610.00
Pack of 500
View alternative products
Save Up to 
»
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
Description
Description
Vented and non-vented
  • Molded of high density polyethylene (HDPE)
  • Designed to prevent misthreading and to provide a secure seal
  • Vented cap has an integral hydrophobic 0.2μm microporous membrane filter vent, allowing consistent gas exchange when cap is in the closed position
Specifications
Specifications
500 Cs.
Sterile
TufRol roller bottle