Learn More
Thermo Scientific™ Nalgene™ Petri Dish
GREENER_CHOICE
Product Code. 11359724
Change view
Click to view available options
Diameter (Metric):
100 mm
Unit Size:
Pack of 60
Product Code. Diameter (Metric) unitSize
11359724 100 mm Pack of 60
1 options
Product Code. 11359724 Supplier Thermo Scientific™ Supplier No. 55000010
Only null left
Add to basket
Add to basket
Shatterproof and reusable
  • Resists cell adhering
  • Stackable for convenience
  • Top and bottom interlocking ridges
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material PMP
Shape Round
Sterility Sterile
Type Petri Dish
Edges Stackable
Quantity Pk, of 10
Height (Metric) 15 mm
Diameter (Metric) 100 mm