missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Centrifuge Bottle Sealing Cap Assembly
GREENER_CHOICE

Product Code. 10192554
Change view
Click to view available options
Includes:
Blue Silicone Gasket
Red Silicone Gasket
Unit Size:
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
10192554 Blue Silicone Gasket Pack of 6
11715233 Red Silicone Gasket Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10192554 Supplier Thermo Scientific™ Supplier No. DS3132BLUE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Includes polypropylene screw cap and shoulder ring, Tefzel™ ETFE plug, and Viton™ fluoroelastomer O-ring to form a leakproof† seal

Seals centrifuge bottle

  • For use with 1L centrifuge bottles
  • Autoclavable
  • Polypropylene screw closure
  • For use at recommended maximum speeds (10,000 x g or greater) without leakage, even when full

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Includes Blue Silicone Gasket
Material Polypropylene
For Use With (Equipment) 1 L PC Nalgene Centrifuge Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.