missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Maxima™ First Strand cDNA Synthesis Kit Reaction Mix (5X)

Product Code. 12938194 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
0.8mL
Unit Size:
800µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12938194 0.8mL 800µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12938194 Supplier Thermo Scientific™ Supplier No. R1362
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ Maxima™ 5X Reaction Mix is a component of the Maxima First Strand cDNA Synthesis Kits for RT-qPCR (K1671/K1672/K1641/K1642) and may be purchased separately. The 5X Reaction Mix is comprised of reaction buffer, dNTPs, oligo (dT)18 and random hexamer primers.

The kit uses Maxima Reverse Transcriptase, an advanced enzyme derived by in vitro evolution of M-MuLV RT. The enzyme features high thermostability, robustness and increased cDNA synthesis rate compared to wild type M-MuLV RT. The contents of the kit include the Maxima Enzyme Mix, 5X Reaction Mix and Water, nuclease-free. Maxima Enzyme Mix contains Maxima Reverse Transcriptase and RiboLock RNase Inhibitor. The recombinant RiboLock RNase Inhibitor effectively protects RNA templates from degradation by RNases A, B and C at temperatures up to 55°C. 5X Reaction Mix contains the remaining reaction components: reaction buffer, dNTPs, oligo (dT)18 and random hexamer primers. Water, nuclease-free is provided for reaction set-up and dilution of sample RNA. The absence of endo-, exodeoxyribonucleases, ribonucleases and phosphatases has been confirmed by appropriate quality tests.

The Thermo Scientific Maxima First Strand cDNA Synthesis Kit for RT-qPCR is a convenient system optimized for cDNA synthesis in 2-step quantitative RT-PCR (RT-qPCR) applications. The kit uses Maxima Reverse Transcriptase (RT), an advanced enzyme derived by in vitro evolution of M-MuLV RT. The enzyme features high thermostability, robustness and increased cDNA synthesis rate compared to wild type M-MuLV RT.
The Maxima First Strand cDNA Synthesis Kit for RT-qPCR is capable of reproducible cDNA synthesis from a wide range of total RNA amounts (1 pg to 5 μg) at elevated temperatures (42 to 65°C).The synthesis reaction can be completed in 15 to 30 minutes. Components of the Maxima First Strand cDNA Synthesis Kit for RT-qPCR are pre-mixed to save time and to reduce the possibility of pipetting errors.
The new Maxima First Strand cDNA Synthesis Kit with dsDNase provides a dramatically simplified workflow that combines genomic DNA elimination and cDNA synthesis into one-tube procedure. The kit contains a novel double-strand specific DNase (dsDNase) engineered to remove contaminating genomic DNA from RNA preps in 2 minutes without damage to quality or quantity of RNA. Highly specific activity towards double-stranded DNA ensures that single-stranded DNA (such as cDNA and primers) is not cleaved and dsDNase treated RNA can be directly added to reverse transcription.
Highlights
• Integrated gDNA removal step
• High yields of full length cDNA up to 20 kb.
• Efficient cDNA synthesis at wide temperature range from 42°C to 65°C.
• Increased synthesis rate - complete cDNA synthesis in 15-30 minutes.
• High sensitivity and specificity.
Applications
• 2-step RT-PCR
• 2-step RT-qPCR
Includes
Maxima First Strand cDNA Synthesis Kit for RT-qPCR contains
• Maxima Enzyme Mix (Maxima Reverse Transcriptase and RiboLock RNase Inhibitor)
• 5X Reaction Mix (reaction buffer, dNTPs, oligo (dT)18 and random hexamer primers)
• nuclease-free water Maxima First Strand cDNA Synthesis Kit with dsDNase contains
• Maxima Enzyme Mix (Maxima Reverse Transcriptase and RiboLock RNase Inhibitor)
• 5X Reaction Mix (reaction buffer, dNTPs, oligo (dT)18 and random hexamer primers)
• dsDNase
• 10X dsDNase Buffer
• nuclease-free water Additional Information about Reaction Components
The recombinant RiboLock RNase Inhibitor, included in Maxima Enzyme Mix, effectively protects RNA templates from degradation by Rnases A, B and C at temperatures up to 55°C.
Oligo(dT)18 and random hexamer primers are supplied with the kit. Random hexamer primers bind non-specifically and are used to synthesize cDNA from all RNAs in a total RNA population. The oligo(dT)18 primer selectively anneals to the 3'-end of poly(A) RNA, synthesizing cDNA only from poly(A)-tailed mRNA. Gene-specific primers may also be used with the kit to prime synthesis from a specified sequence.
Water, nuclease-free is provided for reaction set-up and dilution of sample RNA. The absence of endo-, exodeoxyribonucleases, ribonucleases and phosphatases has been confirmed by appropriate quality tests.
Maxima Universal First Strand cDNA Synthesis Kit
Only available in US and Canada
The Maxima Universal First Strand cDNA Synthesis Kit differs from the Maxima First Strand cDNA Synthesis Kit by having separate kit c

Hazardous:No
>Quality Control:
>•Functionally tested in two-step RT-qPCR using different starting amounts (5 ng to 0.5 fg) of RNA transcript in reverse Transcription reactions followed by amplification with Maxima SYBR Green/ROX qPCR Master Mix.
>•Efficiency is in the range of 90 to 110%, the slope is between -3.09 and -3.58 with a correlation coefficient > 0.99.
> Shipping Condition:Dry Ice
> Storage Condition:-20°C

Two step RT-qPCR.

Order Info

Orders placed Monday through Wednesday ship next day; Thursday and Friday orders ship the following Monday.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 0.8mL

This product or the use of this product is covered by international patent application WO/2009/125006. The purchase of this product includes a non-transferable license to use this product for the purchaser's internal research. All other commercial uses of this product, including without limitation product use for diagnostic purposes, resale of product in the original or any modified form or product use in providing commercial services require a separate license from Fermentas.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.