missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Thermo Scientific™ Lysis Caps, Domed

Product Code. 17829152 Shop All Thermo Scientific Products
This item is not returnable. View return policy
Product Code. 17829152 Supplier Thermo Scientific™ Supplier No. A56895
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Compatible with the lysis tubes in the SureTect Real Time PCR system for pathogen detection, the domed lysis caps enable maximum flexibility for sample processing in your laboratory.

Thermo Scientific™ Lysis Lysis Caps, Domed, strips of 8, are compatible with the 96-well SureTect Lysis Plates as an alternative to Thermo Scientific™ 96-well Piercable Seals. The cap strips have large end tabs for easy strip removal and are compatible for use with the Thermo Scientific™ Capping and Uncapping Tools.

Combined with our comprehensive portfolio of assays and world-class instrumentation, you can perform food safety tests confidently knowing exactly what you are getting – quality combined with simplicity.

We are committed to enabling you to experience the true confidence in your results, right-first time, every time, and the Thermo Scientific™ SureTect™ Real-Time PCR System is continually evolving to support your laboratory and testing needs.

TRUSTED_SUSTAINABILITY

Specifications

Description Lysis Caps, domed
Content And Storage Room Temperature
For Use With (Application) Food & Beverage
Quantity 120
Plate Type Compatible with the 96-well SureTect Lysis Plates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.