Learn More
Thermo Scientific™ Locator™ Cryogenic Rack & Box Systems
Product Code. 10062733 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Locator 4
Locator 6 Plus
Locator 8
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10062733 Locator 4 Each
10328253 Locator 6 Plus Each
10608333 Locator 4 Each
10688493 Locator 8 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10062733 Supplier Thermo Scientific™ Supplier No. HR509X2A70
Includes
Stainless-steel indexed racks that fit into corresponding locations in the vessel. Each rack has individual storage compartments that hold one Thermo Scientific Nalgene™ Cryogenic Storage Box.
Requires
Nalgene Cryogenic Storage Boxes (order separately). Each box has individually numbered storage areas for use with vials sized 2.0mL and smaller.
Inventory system that makes cell sample retrieval quick and easy
Compact design minimizes space requirements while maximizing storage capacity. Suited for both manual and computerized inventory record-keeping methods. Super-insulated vessels have special, narrow-mouth design that conserves liquid nitrogen by minimizing evaporation.
  • Outstanding temperature uniformity: samples are stored below -180°C, even when less than 2 in. (5cm) of liquid nitrogen remains in the vessel
  • Ultrasonic Level Monitor safeguards irreplaceable samples with minimal liquid nitrogen evaporation and conduction
  • Advanced vacuum insulation minimizes liquid nitrogen evaporation and reduces operating costs
  • Optional monitor provides continuous LED readout of liquid nitrogen level; activates visual and audible alarms when level falls below safe range of approximately 1 in. (2.5cm)
  • Optional alarm also alerts operator in the event of excessive liquid nitrogen loss
  • Secure locking hasp prevents unauthorized entry
Locator Jr.
  • Static holding time: 70 days
  • 1600 (100/box)1
  • Evaporation rate: 0.85L/day
Locator 4
  • Static holding time: 112 days
  • 3600 (100/box)2
  • Evaporation rate: 0.99L/day
Locator 8
  • Static holding time: 185 days
  • 1800 (25/box)3
  • Evaporation rate: 0.6L/day
Certifications
CSA, CE
TRUSTED_SUSTAINABILITY
Specifications
Type Cryo Rack
For Use With (Equipment) Locator 4