missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Labtips Pipette Tips
GREENER_CHOICE

Product Code. p-3207576
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
1000 μL
Color:
Blue
Format:
Rack
missing translation for 'unitSize'
Pack of 1800
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Color Format unitSize
12336582 1000 μL Blue Rack Pack of 1800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 12336582 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' 94300230
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Produced from the highest quality virgin polypropylene

  • Double injection molded for maximum sample delivery and best fit for a variety of pipetters
  • Available in 200 and 1000μL including bulk and racked formats

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1000 μL
Sterility Non-sterile
Color Blue
Autoclavable Autoclavable
Format Rack
Tip Style Universal Fit
For Use With (Equipment) LabTip Pipette
Product Line LabTip
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.