Learn More
Thermo Scientific™ High Purity Single Stage Brass Gas Regulators
Product Code. 10351505 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
CGA 320 with Triple Filter
CGA 580 with Triple Filter
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
10351505 CGA 580 with Triple Filter Each
10280005 CGA 320 with Triple Filter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10351505 Supplier Thermo Scientific™ Supplier No. 60180901
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Diffusion-resistant, diaphragm packless outlet valve to maintain system purity
Recommended for applications where precise flow control is not critical
  • For non-corrosive analytical and process applications
  • Includes a built-in Smart Purifier Base and Cartridge ensuring 99.99999% gas purity
  • Specially designed, convoluted, stainless steel diaphragm provides good regulating performance and maximum purity integrity without the need for a soft seal
  • Capable of withstanding an internal vacuum
  • All regulators supplied with 1/8 in. stainless steel output adaptor
  • Built in check valve allows for cylinder change without contaminating the gas line
TRUSTED_SUSTAINABILITY
Specifications
Range 0 to 200 psig
Type CGA 580 with Triple Filter