missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Whole Blood RNA Purification Mini Kit

Product Code. 11511655 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 Preps
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11511655 50 Preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11511655 Supplier Thermo Scientific™ Supplier No. K0761
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Lysis Buffer, Wash Buffer WB 1 (concentrated), Wash Buffer 2 (concentrated), Water, nuclease-free, GeneJET™ RNA Purification Columns (pre-assembled with collection tubes), Collection tubes (2mL), Collection tubes (1.5mL), Detailed Protocol

Designed for rapid and efficient purification of high quality total RNA from whole blood and related body fluids

Thermo Scientific GeneJET Whole Blood RNA Purification Mini Kit is designed for rapid and efficient purification of high quality total RNA from whole blood and related body fluids.

The kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The standard procedure takes less than 15 minutes. The purified high quality RNA can be used in a wide range of downstream applications, such as RT-PCR, RT-qPCR, Northern blotting, and other RNA-based analysis.

Highlights

  • Efficient – up to 1.8 μg of total RNA from 500 μL of human blood
  • Convenient – spin columns are capped and assembled with collection tubes
  • Pure – RNA pure from enzyme inhibitors, proteins, ready to use in all demanding applications. Validated in cDNA synthesis and quantitative RT-PCR
  • Fast and simple – procedure takes less than 15 minutes and does not involve organic extraction or differential RBC lysis
  • Compatible – suitable for vide variety of samples - human and animal blood treated with EDTA, citrate or heparin, buffy coat, bone marrow, and related body fluids

Applications

  • Fast extraction of high purity RNA suitable for all conventional molecular biology procedures, including:
  • RT-PCR
  • RT-qPCR
  • Northern blotting
  • Nuclease protection assaysncludes Lysis Buffer Wash Buffer WB 1 (concentrated) Wash Buffer 2 (concentrated) Water, nuclease-free GeneJET RNA Purification Columns (pre-assembled with collection tubes) Collection tubes, 2 mL Collection tubes, 1.5 mL Detailed Protocol
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 40 mL Lysis Buffer; store at room temperature
• 40 mL Wash Buffer WB1 (concentrated); room temperature
• 23 mL Wash Buffer 2 (concentrated); room temperature
• 30 mL Nuclease-free Water; room temperature
• 50 GeneJET RNA Spin Columns with Collection Tubes; For better long-term performance, it is recommended to store the purification columns at 2°C-8°C.
• 50 Collection Tubes (2 mL); room temperature
• 50 Collection Tubes (1.5 mL); room temperature
Isolation Technology Silica Spin Column
Purification Time 15 min. (After sample homogenization)
Elution Volume 50 μL
Sample Type Blood, Bone Marrow, Buffy Coat
Final Product Type Total RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Quantity 50 Preps
Scale Mini
Shipping Condition Room Temperature
Starting Material Amount Blood: 50–500 μL, Bone Marrow: 50–500 μL, Buffy Coat: 500 μL
Yield Up to 11 μg
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.