Learn More
Thermo Scientific™ Wash Buffer 2 for MagJET Plasmid DNA Kit (concentrated)
Shop All Thermo Scientific ProductsDescription
The Thermo Scientific™ MagJET™ Plasmid DNA Kit is designed for fast and efficient purification of plasmid DNA from overnight cultures of E. coli cultures. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.
Specifications
Specifications
| Buffer | Wash Buffer |
| Format | Kit |
| Product Line | MagJET |
| Product Type | Wash Buffer |
| Quantity | 60 mL |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.