missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lysis Buffer for MagJET RNA Kit

Product Code. 13525190 Shop All Thermo Scientific Products
Click to view available options
Quantity:
200 mL
Unit Size:
200mL
This item is not returnable. View return policy

Product Code. 13525190

Brand: Thermo Scientific™ R2302

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Generate sample lysate in order to isolate total RNA. Thermo Scientific™ MagJET™ Lysate Buffer is a component of the MagJET™ RNA Kits (K2731/2732) and may be purchased separately when additional solution is needed. It is provided in 200mL bottles.

The Thermo Scientific™ MagJET™ RNA Kit is designed for fast and efficient purification of total RNA from cultured mammalian cells, tissues, bacteria, and yeast. The kit utilizes paramagnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality RNA is free of proteins, nucleases and other contaminants or inhibitors.

TRUSTED_SUSTAINABILITY

Specifications

Buffer Lysis Buffer
Product Line MagJET
Quantity 200 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.