Learn More
Thermo Scientific™ Binding Buffer for MagJET Blood gDNA Kit (concentrated)
Shop All Thermo Scientific ProductsDescription
The Thermo Scientific™ MagJET™ Whole Blood Genomic DNA Kit is designed for fast and efficient purification of genomic DNA from fresh or frozen whole blood treated with EDTA or citrate, as well as blood fractions. The kit utilizes paramagnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.
Specifications
Specifications
| Buffer | Binding Buffer |
| Format | Kit |
| For Use With (Equipment) | MagJET Blood gDNA Kit |
| Product Line | MagJET |
| Product Type | Binding Buffer |
| Quantity | 23 mL |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.