Learn More
Epredia™ Disposable Blade Carriers
Product Code. 12063119 Shop All Epredia Products
Change view
Click to view available options
For Use With (Equipment):
Long Stroke Units
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
12063119 Long Stroke Units Each
1 options
Product Code. 12063119 Supplier Epredia™ Supplier No. 705830
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Utilize a quick clamp system that accepts low- and high-profile blades
Updated models (long stroke and version 2 bar guideways) include removable angle adjustment and blade clamp levers.
Updated Disposable Blade Carriers
  • Blade angle and blade adjustment levers are removable to help protect user settings
  • Concentric lever locks onto the dovetail to help reduce accidental movement of the holder
  • Only compatible with newer HM 325, HM 355S, and HM 340E models which have serial numbers above 34000
Standard Disposable Blade Carriers
  • Quick clamp system for high- and low-profile blades
  • Clearance angle is adjustable
  • Upper part moves laterally without opening the blade clamp
  • Moveable knife guard covers the blade entirely in any position
  • Complete with knife carrier base
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Long Stroke Units