missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CycleBox Recycle PP#5 Pipette Tip Racks, No tips

Product Code. 17805746 Shop All MBP Molecular BioProducts Products
Change view
Click to view available options
Volume (Metric):
96 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric)
17805746 96 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17805746 Supplier Thermo Scientific™ Supplier No. CYCLEBOX
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ CycleBox Recycle Kit mail-back program for recycling PP#5 pipette tip racks; no tips. Fill box adhering to acceptable materials. Securly seal box top and bottom. Request collection by sending an email and fill out waste transfer note and arrange a pickup within 3-5 working days.

Thermo Scientific™ CycleBox Recycle Kit mail-back program provides a simple and effective solution for recycling your ThermoScientific branded PP#5 nonhazardous pipette tip racks; Tips excluded from program. Fill box adhering to list of acceptable materials. Securely seal box top and bottom. Request collection by sending an email, fill out waste transfer note and arrange a pickup within 3-5 working days. Material is delivered to facility that converts it back into pellet format so it can be used again.

  • Recycle nonhazardous pipette tip racks with CycleBox.
  • For use with any ThermoFisher Scientific branded PP#5 pipette tip boxes.
  • A few simple steps needed to recycle PP#5.
  • Order Cyclebox and fill box with racks only, no tips, adhering to list of acceptable materials.
    • If delivered material is contaminated, a non-conformance procedure may be initiated with potential surcharges, and the recycling box will be returned to you.
  • Once full, seal the box with adhesive tape, close box and securely tape bottom and top. Request a collection by sending an email with Subject line ""ThermoFisher - Pipette Tip Box Recycling"" to uk.veolia.waste.procycle.mailbox@veolia.com
  • Upon email receipt a response provided with following information: Waste Transfer Note, complete Section A & B. Email the signed Waste Transfer Note, along with attached photo of the box and contents to uk.veolia.waste.procycle.mailbox@veolia.com to finalize the pickup request.
  • Once information is validated, pickup arragement coordinated within 3-5 working days. The courier will bring a shipping label and attach it to the box during pickup. The box must be ready to be handed over to the courier on arrival without waiting time. A failed collection will incur a charge for any subsequent attempts.
  • Material is delivered to facility, converted back into pellet format so it can be used again.
TRUSTED_SUSTAINABILITY

Specifications

Description Recycle PP#5 pipette tip racks
Volume (Metric) 96 L
Features 400 x 400 x 600 mm rack
Format Box
Quantity 1 Recycle Box per pack
Shipping Condition Ambient
Material Cardboard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.