Learn More
Thermo Scientific™ CryoMed™ Thermocouple Sensors
Product Code. 10664325 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Ribbon Type
Thermocouple Sensor
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10664325 Ribbon Type Each
10001164 Thermocouple Sensor Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10664325 Supplier Thermo Scientific™ Supplier No. 4000393
 Restricted Item
In Stock
Add to basket
Add to basket
Sample thermocouple sensors
Thermocouple sensors for cryo tubes, sheath straw and vial samples.
  • Include male plug
For controlled-rate freezing applications
TRUSTED_SUSTAINABILITY
Specifications
Type Ribbon Type
For Use With (Equipment) Cyrogenic Freezer
Plug Type Male
Holds Cryo Tube, Sheath Straw, Vial