Learn More
Thermo Scientific™ Clear Seal Heat Sealing Film
Product Code. 10187604
Click to view available options
:
For ALPS 25 Automated Laboratory Plate Sealer; Sheets
Product Code. 10187604
Brand: Thermo Scientific™ AB1797
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ideal for applications requiring pierceable non-conductive seal material
  • Suitable for PCR applications with screw- or clip-down heated lid thermal cyclers only
  • Seals polypropylene, polystyrene, polyethylene and Diamond plates
  • Easily pierced and peelable by hand from 96-well plates
  • Temperature range: -80°C to +80°C