missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Castaloy™ Three-Prong Extension Clamps

Hold irregular shapes at any angle, surely and safely.

Brand:  Fisherbrand™

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 11343982

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Sturdy extension clamps allow you to hold objects with irregular shapes at any angle, safely and securely.

 

  • Two separate adjustment screws ensure even, precise pressure on glass and minimize chance of breakage
  • Clamps are easily adjustable to vary holding angle, distance from stand

 

Specifications

Specifications

1 Ea.
Fiberglass (Sleeve), Vinyl (Sleeve)
6.5 in.
Extension arm O.D.: 0.31 in. (0.8cm)
1 in.
Extension Clamp
Fiberglass and Vinyl sleeve
17 cm
2.5 cm
3-Prong
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers