missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BioBasic™ Phenyl HPLC Columns

Product Code. p-3616096 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
250 mm
Diameter (Metric):
4.6 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10322935

Brand: Thermo Scientific™ 72405254630

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Alternative selectivity of peptides and proteins

  • 300Å pore size for maximum performance with biomolecules
  • Lower carbon load for optimal retention of larger peptides and proteins
  • Offers different selectivity to alkyl chain chemistires for critical separations
  • Excellent for LC/MS separations

Order Info

Other column dimensions are available. Please call Customer Service for more information.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 250 mm
Particle Size 5 μm
Pore Size 300 Å
USP Type L11
Stationary Phase Phenyl (Ph)
Product Line BioBasic
Packing Material Silica
Carbon Load 3%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.