missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- 300Å pore size for maximum performance with biomolecules
- Lower carbon load for optimal retention of larger peptides and proteins
- Offers different selectivity to alkyl chain chemistires for critical separations
- Excellent for LC/MS separations
Order Info
Other column dimensions are available. Please call Customer Service for more information.
Specifications
Specifications
| Diameter (Metric) | 4.6 mm |
| Length (Metric) | 250 mm |
| Particle Size | 5 μm |
| Pore Size | 300 Å |
| USP Type | L11 |
| Stationary Phase | Phenyl (Ph) |
| Product Line | BioBasic |
| Packing Material | Silica |
| Carbon Load | 3% |
| Column Type | Reversed Phase |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?