missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BioBasic™ Phenyl Drop-in Guard Cartridges

Product Code. p-3647694 Shop All Thermo Scientific Products
Click to view available options
Diameter (Metric) Inner:
1 mm
Unit Size:
Pack of 4
This item is not returnable. View return policy

Product Code. 10310915

Brand: Thermo Scientific™ 72405011001

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Requires

Thermo Scientific UNIGUARD™ direct connection or stand alone holder

Offer convenience, economy and effective protection for extending column lifetime

  • The 10mm design offers maximum protection with minimal increase in retention
  • Fit Thermo Scientific UNIGUARD direct connection and stand alone holders
  • Available in packs of 4

Features of BioBasic Phenyl Solid Phase

  • 300Å pore size for maximum performance with biomolecules
  • Lower carbon load for optimal retention of larger peptides and proteins
  • Offers different selectivity to alkyl chain chemistires for critical separations
  • Excellent for LC/MS separations

Order Info

Thermo Scientific UNIGUARD™ direct connection or stand alone holder

TRUSTED_SUSTAINABILITY

Specifications

Particle Size 5 μm
Length (Metric) 10 mm
Diameter (Metric) Inner 1 mm
Stationary Phase Phenyl
Product Line BioBasic
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.