missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BetaBasic™ 4 HPLC Columns, 5μm Particle Size

Product Code. p-3136301 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
50 mm
250 mm
Diameter (Metric):
2.1 mm
4.6 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10087554

Brand: Thermo Scientific™ 71605052130

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

A good first choice for compounds with poor peak symmetry on other columns

  • 150Å pore size for small molecules, peptides and protein digests
  • Superb pH stability

Order Info

Other column dimensions are also available. Please call Customer Service for more information.

TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 2.1 mm
Length (Metric) 50 mm
Particle Size 5 μm
Pore Size 150 Å
USP Type L26
Stationary Phase C4
Product Line Beta
Packing Material Silica
Carbon Load 6%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.