missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BetaBasic™ 4 HPLC Columns, 5μm Particle Size
Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
50 mm
250 mm
Diameter (Metric):
2.1 mm
4.6 mm
Unit Size:
Each
Description
- 150Å pore size for small molecules, peptides and protein digests
- Superb pH stability
Order Info
Other column dimensions are also available. Please call Customer Service for more information.
Specifications
Specifications
| Diameter (Metric) | 4.6 mm |
| Length (Metric) | 250 mm |
| Particle Size | 5 μm |
| Pore Size | 150 Å |
| USP Type | L26 |
| Stationary Phase | C4 |
| Packing Material | Silica |
| Carbon Load | 6% |
| Column Type | Reversed Phase |
| Endcapped | Yes |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction