missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Thermo Scientific Barnstead; For Use with: Nanopure Quad, E-pure; Cartridge: Ultrapure Ion Exchange
Pretreatment/Prefilter, Thin Film
- Extruded carbon cartridge (with 5μm prefilter) extends life of 09-050-37 membrane by removing chlorine from feedwater
- Capacity: 760 grains as NaCl to 1megohm endpoint
- Can also be used with E-pure™ systems and Nanopure™ systems
Specifications
Specifications
| Capacity | 760 grains |
| Purification Method | Ultrapure |
| For Use With (Application) | Spring ion exchange |
| For Use With (Equipment) | Nanopure Quad and E-Pure System, Spring Ion Exchange |
| Type | Cartridge |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?