Learn More
Thermo Scientific™ Barnstead™ Easypure™ II Compact Ultrapure Water System Filters and Cartridges
Brand: Thermo Scientific™ D50229
Code : 03
Additional Details : Weight : 0.72000kg
Description
- Easypure™ II Systems require pretreated feedwater
- From the chart, select appropriate cartridges based on application and feedwater type
Order Info
Cartridges and filters must be ordered separately
Specifications
Organicfree/organic Removal | |
Cartridge and Filter | |
60 μm |
Easypure II Water Purification System, Organic Free/Organic Removal | |
RO or Distilled | |
Ultrapure Mixed Bed |