missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Acclaim™ PepMap™100 Fusica™ Capillary Columns, 5μm Particle Size
Feature high loading capacity for exceptional suitability in the analysis of low abundant peptides in complex proteomics samples
Brand: Thermo Scientific™ 164901
Code : NEW
Additional Details : Weight : 0.50000kg
Description
- High resolution in protein identification, biomarker discovery, and systems biology
- Particular loadability allows hghest sensitivity in LC/MS
- Designed for TFA-free LC-MS, minimizing ion-suppression effects
- Optimized for coupling to ESI/MS and MALDI-MS
- Highest column-to-column reproducibility
- Easy-to-use, cutting-edge miniaturized HPLC
- Inside diameter 300μm
Specifications
Capillary Column | |
50 mm | |
2 to 8 | |
450 m²/g | |
C18 | |
Silica | |
Reversed Phase | |
Acclaim PepMap C18 100 A nanoViper HPLC Column |
0.3 mm | |
5 μm | |
100 Å | |
L1 | |
nanoLC | |
15% | |
Reversed Phase |