Learn More
Thermo Scientific™ ABgene™ Thermo-Mat™ Sealing Mats
Description
Thermo Scientific 96-well Sealing Mats are designed for 96-well storage plates and are available in either piercable or autoclavable formats. The round or square capped mats are made of thermoplastic elastomer and are suitable for long-term storage.
Highlights
- Round or square capped mats for sealing
- Alphanumeric grid-referencing
- Molded from a proprietary thermoplastic elastomer for a flexible design
- Suitable for long term storage
- Available in autoclavable or piercable formats
Pierceable version:
- Solvent resistant (including DMSO, Ethanol, and Isopropanol)
- Suitable for UV Irradiation and Gamma Irradiation
- Requires 6N for piercing
- Temperature Range: -20°C to +55°C
Autoclavable version:
- Suitable for UV Irradiation and Gamma Irradiation
- Temperature Range: -20°C to +130°C
Hazardous: No
Storage Condition: Ambient
Certifications
Certified free from DNase, RNase and human genomic DNA.
Specifications
Specifications
| Certifications/Compliance | Certified free from DNase, RNase and Human Genomic DNA |
| Material | TPE |
| Autoclavable | Autoclavable |
| No. of Wells | 96 |
| No. per Pack | 5 |
| Temperature Range (English) | -4°F to 266°F |
| Product Type | Sealing Mat |
| Temperature Range (Metric) | -20°C to +130°C |
| Plate Compatibility | 96-well PCR Plate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.