missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ABgene™ Thermo-Fast™ 96 PCR Detection Plate MKIII

Product Code. 10150883 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Clear
White
Barcode:
Includes Barcode
No Barcode
Unit Size:
Pack of 25
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Barcode unitSize
10150883 Clear No Barcode Pack of 25
10607513 White No Barcode Pack of 25
10576910 White Includes Barcode Pack of 25
10231310 Clear Includes Barcode Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10150883 Supplier Thermo Scientific™ Supplier No. AB1900

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

96-well low-profile semi-skirted PCR plate for the fast block of Life Technologies PCR and qPCR thermal cyclers. Supplied with barcodes for sample and plate traceability.

The Thermo Scientific 96-Well Semi-Skirted, Fast Block, Barcoded Plates is compatible with all ABI Fast block machines, including Life Technologies PCR and qPCR thermal cyclers. These plates feature a raised deck to prevent spills and a cut corner at the A1 location.
The 96-Well Semi-Skirted, Fast Block, Barcoded Plates are available in white plates for qPCR applications and have clear wells for maximum sample visibility.

Highlights

  • Directly compatible with all Applied Biosystems thermal cycler fast blocks with no adapters necessary
  • Raised deck of plate to aid in spill prevention
  • 0.1 mL maximum well volume (when used with adhesive and heat seals)
  • Clear well for sample visibility
  • Uniform, thin well walls deliver maximum and consistent heat transfer
  • White plates are optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
  • Cut Corner: A1
Applications
  • PCR
  • qPCR/Real-Time PCR
The barcodes are the industry standard code 128. The label itself can withstand temperatures from -196°C up to 120°C to ensure that the barcodes stay on the plate throughout your entire workflow. It also has a special coating to virtually eliminate any 'rubbing off' of the code during handling. The dual barcodes are located on the opposite numeric and opposite alpha sides of the plate.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance DNA, RNase, Endotoxin, DNase-free
Format Standard
Array 8 x 12
Color Clear
Material Polypropylene
No. of Wells 96
For Use With (Application) PCR, qPCR, RT-PCR
Quantity 25/Pk.
Rimmed Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.