missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SuperPlate PCR Plates, 96-well

Product Code. 10241310
Change view
Click to view available options
Color:
Blue
Clear
White
Yellow
Skirt Style:
Full
Semi
Product Type:
Raised Deck
Barcode:
Includes Barcode
No Barcode
Unit Size:
Pack of 25
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Skirt Style Product Type Barcode unitSize
10241310 Clear Semi Raised Deck Includes Barcode Pack of 25
10124183 Clear - - No Barcode Pack of 25
10183653 White - - No Barcode Pack of 25
10657513 Blue Full Raised Deck No Barcode Pack of 25
11802133 Yellow Full Raised Deck No Barcode Pack of 25
10681331 Clear - - - Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10241310 Supplier Thermo Scientific™ Supplier No. BC2100

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

96-well PCR plates with a flat deck for improved sealing in PCR and qPCR applications.

The Thermo Scientific 96-Well Semi-Skirted Flat Deck PCR Plates are compatible with most thermal cyclers and ABI platforms, including sequencers, with no adapters necessary. The Thermo Scientific PCR plastics are designed, manufactured, and tested to ensure PCR performance. The plates have 0.3 mL maximum well volume when used with adhesive and heat seals.
The flat plate deck design ensures easy handling, and the plates feature a raised rim to prevent evaporation. The 96-Well Semi-Skirted Flat Deck PCR Plates have a cut corner at the A12 location and have thin well walls for consistent heat transfer.
Highlights
• Directly compatible with all major major thermal cyclers, including ABI platforms and sequencers with no adapters necessary
• Flat plate deck facilitates sealing and handling
• Available as SuperPlate providing 4x more rigidity for superior robotic handling
• Raised rim design around each well enables secure sealing and safeguards against evaporation
• Uniform, thin well walls deliver maximum and consistent heat transfer
• Black lettered versions feature black alpha-numeric lettering to allow for easy well identification
• White plates are optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
• Cut corner: A12
Applications
• PCR
• qPCR/Real-Time PCR
• Sequencing

Hazardous:No
>Storage Condition:Ambient

TRUSTED_SUSTAINABILITY

Specifications

Barcode Includes Barcode
Certifications/Compliance DNA, DNase, RNase-free
Format Standard
Array 8 x 12
Color Clear
Product Type Raised Deck
Skirt Style Semi
Material Polypropylene
Volume (Metric) Well 300 μL
No. of Wells 96
For Use With (Application) For improved sealing in PCR and qPCR applications
Quantity 25/Pk.
Rimmed Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.