Learn More
Thermo Scientific™ 48-Well Semi-Skirted Plates
Product Code. 10640201
Change view
Click to view available options
Color:
Purple
White
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10640201 Purple Pack of 50
12687416 White Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10640201 Supplier Thermo Scientific™ Supplier No. AB0648/P
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
48-well semi-skirted plate for PCR and qPCR applications.
The Thermo Scientific 48-well semi-skirted plate for PCR and qPCR applications comes in a convenient format, which allows for lower sample numbers. The semi skirt makes the plate more rigid and allots space for labeling and barcoding. Available in assorted colors, it's compatible with standard multichannel pipettes and automated liquid handling systems.The plate is sealable when used with adhesive films, foils or cap strips.
Highlights
• Convenient format for lower sample numbers
• Semi-skirt adds rigidity and allows for labeling or barcoding
• Sealable using adhesive films and foils and flat, ultra clear and domed cap strips
• 0.3 mL maximum well volume (when used with adhesive and heat seals)
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 6 x 8
Color Purple
Material Virgin Polypropylene
No. of Wells 48