missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 384-well Microarray Plate

Product Code. 10669973
Change view
Click to view available options
Color:
Natural
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10669973 Natural Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10669973 Supplier Thermo Scientific™ Supplier No. AB1055

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

384-well microarray plate designed for low volume sample storage.

Thermo Scientific Storage and Assay Plates are manufactured and in a class 100,000 clean room environment to ensure that each plate is contamination free. Plates are made using virgin medical grade polypropylene and have highly polished surfaces making them low binding and chemically resistant. Rigid side walls and industry standard footprint make plates suitable for automation and robotic handling, as well as for low throughput applications.
Highlights
• Designed for optimal sample recovery
• Each well specifically designed with an independent sealing rim to prevent cross-contamination
• Ideally suited for use with the widest range of sealing options
• Improved sample recovery and decreased dead volume
• Conical bottom (70 μL maximum volume) wells

• Compatible Sealing Film: Adhesive, Heat
• Hazardous: No
• Max Centrifugal Force: 5000 g
• Max Vol (Cap Seal): n/a
• Max Vol (Film Seal): 70 μL (AB-1055) or 55 μL (AB-1056)
• Max Vol (Mat Seal): 30 μL (AB-1055) or 25 μL (AB-1056)
• Storage Condition: Ambient

Certifications

Certified free from human DNA, RNases, and DNases.

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Array 16 x 24
Color Natural
Material Polypropylene
No. of Wells 384
Well Shape Round
For Use With (Application) For use with the widest range of sealing options
Max. RCF 5000 x g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.