missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Thermo Scientific™ 2D Tank Liner, ASI™ 26 film

Single-use tank liners reduce cleaning steps common to re-usable tanks.

Brand:  Thermo Scientific™ B100038-I

Additional Details : Weight : 0.45000kg

 View more versions of this product

Product Code. 17880777

  • £300.80 / Each
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MAP4K4 (HGK) is a serine⁄threonine kinase which plays an important role in stress-activated MAPK core signaling pathways.
Specifications

Specifications

130 L
Store at room temperature.
20 per case
Irradiated
HyPerforma™
ASI 26
Polyethylene
45.7 cm x 76.2 cm (18 in. x 30 in.)
SV30112
Room Temperature
0 Ports
No Tubing
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers