missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Thermo Scientific™ 2D Tank Liner, ASI™ 26 film

Product Code. 17830787 Shop All Thermo Scientific Products
Click to view available options
Capacity (Metric):
50 L
200 L
340 L
400 L
560 L
Tubing Style:
No Tubing
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17830787

Brand: Thermo Scientific™ B100049I

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Single-use tank liners reduce cleaning steps common to re-usable tanks.

The Thermo Scientific™ 2D Tank Liner System without bottom drain is composed of Thermo Scientific™ ASI™ 26 Polyethylene Film, a durable 5.0 mL film, with dimensions optimized for Thermo Scientific™ drums and commonly-used, industry standard cylindrical tanks. Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.

  • Minimizes bioburden due to construction under sterile cGMP conditions
  • Eliminates post-use cleaning steps required with reusable containers, helping to reduce cross-contamination risks and reduce cycle times
  • Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 340 L
Material Polyethylene
Content And Storage Store at room temperature.
Dimensions 70 cm x 122 cm (24 in. x 48 in.)
Quantity 340 L, 35 per case
For Use With (Equipment) SV30114
Sterilization Method Irradiated
Shipping Condition Room Temperature
Product Line HyPerforma
Port Detail 0 Ports
Film Description ASI 26
Tubing Style No Tubing
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.