missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Abgene™ 96 Well 2.2mL Polypropylene DeepWell™ Sample Processing & Storage Plate
GREENER_CHOICE

2.2 mL deepwell storage plates for the culture or storage of large samples.

Brand:  Thermo Scientific™ AB-0788

214.61 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11897612

  • £364.00 / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Abgene™ polypropylene microplates and deepwell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use. Abgene microtiter plates are manufactured to exacting specifications in our Class 100,000 clean room ISO 9001 conditions using high quality medical grade virgin polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and deepwell plates are compatible with a variety of automated liquid handling for high throughput workflows. The polypropylene storage plates are offered with spindle, pyramidal, U and V bottom wells along with a wide range of volumes to accommodate most applications.

Highlights
  • Designed for optimal sample recovery
  • Each square well is specifically designed with an independent sealing rim to prevent cross-contamination
  • U-bottom (AB-0661) to aid resuspension or conical bottom (AB-0932) for ease of sample retrieval
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Custom Barcoding Available
96-well
8 x 12
Bulk
Abgene
96-well plate
Polypropylene
96
Cleanroom
Square, Round Bottom (U-well)
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
50
50/Cs.
Yes
ANSI/ SLAS Standard
Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
96-well Polypropylene Sample Processing & Storage Plates
Autoclavable
Low binding PP
Natural
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
2200 μL
1900 μL
Non-treated
Yes
5000 x g
50
For Research Use Only. Not for use in diagnostic procedures.
PCR Ready
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers