Learn More
Thermo Scientific™ Tubes, 0.2 mL, flat cap, red
Description
0.2 mL Individual Tubes with integral “snap shut” caps are suitable for 0.2 mL thermal cycler blocks. They have a 0.25 mL maximum volume when closed. These individual tubes feature thin-wall design and are made from virgin polypropylene. The tubes are cleanroom produced, ensuring that they are free from contaminants that could affect experimental results. They are available in multiple colors with flat or domed caps.
The Thermo Scientific 0.2 mL Individual Tubes with integral ”snap shut“ caps are suitable for 0.2 mL thermal cycler blocks. They have a 0.25 mL maximum volume when closed. These individual tubes feature thin-wall design and are made from virgin polypropylene. The tubes are cleanroom produced, ensuring that they are free from contaminants that could affect experimental results. They are available in multiple colors with flat or domed caps.
Highlights
• Suitable for 0.2 mL thermal cycler blocks
• 0.25 mL maximum tube capacity when closed
•
Integral 'snap shut' cap
• Available with flat or domed caps
Applications
• PCR
• Sample storage
Hazardous:No
>Storage Condition:Ambient
PCR; Sample storage
Certifications
Certified free from human DNA, RNases, and DNases
Specifications
Specifications
| Certifications/Compliance | Certified DNA/RNase Free |
| Color | Red |
| Volume (Metric) Working | 0.2 mL |
| Volume (Metric) | 0.25 mL |
| Quantity | 1000 tubes |
| For Use With (Equipment) | Thermal Cyclers |
| Material | Polypropylene |
| Reaction Speed | Standard |
| Closure Type | Flat Cap |
| Format | 1,000 Tubes |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.