missing translation for 'onlineSavingsMsg'
Learn More

Thermit Powder, Pure, Fisher Chemical

Product Code. 10204120 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10204120 500 g Glass Jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10204120 Supplier Fisher Chemical Supplier No. T/0977/53
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Silver
Physical Form Powder/Solid
Quantity 500 g
MDL Number 148616
Packaging Glass Jar
compliance-icons
Product Identifier
  • Thermit powder
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Substance which, in contact with water, emits flammable gases Category 2
Hazard Statement
  • H228-Flammable solid.
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
  • P231+P232-Handle under inert gas. Protect from moisture.
Supplemental information
  • MIXTURE LIST-Contain: Aluminum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.