missing translation for 'onlineSavingsMsg'
Learn More

Interscience™ Thermal Printer

Product Code. 16385695
Click to view available options
Color:
Black
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16385695

Brand: Interscience™ 410150

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Thermal printer with multiple outputs.

  • Contains RS 232 and USB outputs

Specifications

Product Type Printer
Color Black
Display Type None
For Use With (Equipment) DiluFlow, DilluFlow Pro and Elite, Flexipump Pro
Print Technology Thermal Transfer
For Use With (Application) Printing
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.