Learn More
LTE Scientific™ Thermal Data Printer
Product Code. p-9468005
Click to view available options
For Use With (Equipment):
LTE Touchclave K and F Autoclaves
LTE Touchclave R Autoclaves
Unit Size:
Each
Product Code. 12735385
Brand: LTE Scientific™ TC/TDP/01
Only null left
Add to basket
Add to basket
Thermal Data Printer for LTE Touchlave models.
  • Thermal data printer
  • Prints all cycle data as cycle progresses
  • Printout includes any faults/alerts
  • Provides printed validation of load success
Specifications
For Use With (Equipment) LTE Touchclave K and F Autoclaves
Model TC/TDP/01
Product Type Thermal Data Printer
Sterility Non-sterile