Learn More
LTE Scientific™ Thermal Data Printer
Product Code. 12735385
Change view
Click to view available options
For Use With (Equipment):
LTE Touchclave K and F Autoclaves
LTE Touchclave R Autoclaves
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
12735385 LTE Touchclave K and F Autoclaves Each
12735405 LTE Touchclave R Autoclaves Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12735385 Supplier LTE Scientific™ Supplier No. TC/TDP/01
Thermal Data Printer for LTE Touchlave models.
  • Thermal data printer
  • Prints all cycle data as cycle progresses
  • Printout includes any faults/alerts
  • Provides printed validation of load success
Specifications
For Use With (Equipment) LTE Touchclave K and F Autoclaves
Model TC/TDP/01
Product Type Thermal Data Printer
Sterility Non-sterile