missing translation for 'onlineSavingsMsg'
Learn More

Julabo™ Thermal Bath Fluids

Used for temperature control applications. Julabo™ Thermal Bath Fluids offer safe and reliable operation.

Brand:  Julabo™ 8940111

Code : 50

Additional Details : Weight : 5.20000kg


 View more versions of this product

Product Code. 11924738

  • £454.00 / 5 L
Estimated Shipment: 29-05-2024
to see stock.
Add to basket

Description

Description

  • For closed systems like the Presto or Forte
  • Has wide temperature ranges
  • Low viscosity
  • High stability
  • Good heat conductivity
  • Minimum odor
  • Low corrosion tendency
  • Low toxicity
  • Long shelf life
Specifications

Specifications

Clear
50°C to 350°C
Thermal Bath
H350
450°C
Highly dynamic temperature control systems
Thermal Bath Fluid
Drain tap
5 L
48.3mm2/s at 20°C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • thermal H350

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Aspiration toxicity Category 1

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H413-May cause long lasting harmful effects to aquatic life.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : dibenzyltouene

SDS
Documents

Documents

Certificates
Special Offers

Special Offers