missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TheraPure™ PAC-rA Phosphoramidite, serum vial bottle

Product Code. 17907182 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17907182 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17907182 Supplier Thermo Scientific™ Supplier No. 27180302

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PAC-rA CEP is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with Phenoxyacetyl (PAC) base protection, for use in oligonucleotide synthesis.

PAC-rA CEP is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with Phenoxyacetyl (PAC) base protection, for use in oligonucleotide synthesis. The UltraMILD monomers - phenoxyacetyl (PAC)-dA, acetyl (Ac)-dC, and iso-propyl-phenoxyacetyl (iPr-PAC)-dG - were developed to alleviate the problem of modifiers and labels not withstanding prolonged exposure to strongly alkaline deprotection conditions. Cleavage and deprotection can be achieved in 4 hours at room temperature with 0.05M potassium carbonate in methanol. The TheraPure trademark denotes our highest level of phosphoramidite purity, developed with the greatest stringency of high definition analytics and tightly controlled impurity profiles and, therefore, ideal for any therapeutic application. This product is supplied in a 20 mL Amber bottle with a Serum Vial neck style, which is compatible with most ABI Systems|AKTA Systems of oligonucleotide synthesizers.

Features:
Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-N6-phenoxyacetyl-2'-O-TBDMSi-adenosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
Molecular formula: C54H68N7O9PSi
Base: rA
Base Protection: PAC
Formula weight: 1018.2 g/mol
CAS number: 121058-86-4
HPLC Purity: ≥99%
31P NMR Purity: ≥99%; Sum of non-primary peaks in 140-152 ppm range: ≤0.5%; No single PIII impurity (140-152 ppm) >0.3%

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) AKTA Synthesizers, Applied Biosystems Synthesizers
Product Type PAC-rA CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Adenosine (A)
Reagent Features Fast deprotection
Shipping Condition Room Temperature
Purity 99%
Quantity 1 g
Reagent Type Phosphoramidite
Grade TheraPure™
Nucleic Acid Type RNA
Product Line TheraPure
Form Powder
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.