missing translation for 'onlineSavingsMsg'
Learn More

Theophylline, TRC

Product Code. 18042020
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18042020 100 mg 100mg
18052210 5 g 5g
18011230 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18042020 Supplier Toronto Research Chemicals Supplier No. TRCT343850100MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-55-9
Molecular Formula C7 H8 N4 O2
Molecular Weight (g/mol) 180.16
Synonym 1,3-Dimethyl-3,7-dihydro-1H-purine-2,6-dione, Theophylline, Diprophylline Imp. B (EP), Pentoxifylline Imp. C (EP), Dimenhydrinate Imp. A (EP), Caffeine Imp. A (EP), 1H-Purine-2,6-dione, 3,9-dihydro-1,3-dimethyl-, 1H-Purine-2,6-dione, 3,7-dihydro-1,3-dimethyl- (9CI), Theophylline (8CI), 1,3-Dimethyl-1H-purine-2,6(3H,7H)-dione, 1,3-Dimethylxanthine, 3,9-Dihydro-1,3-dimethyl-1H-purine-2,6-dione, Accurbron, Aerobin, Aerolate, Afonilm, Apnecut, Armophylline, Asmax, Austyn, Bilordyl, Brochoretard, Bronkodyl, Cetraphylline, Chronophyllin, Constant T, Controfilina 200, Diffumal, Duraphyl, Duraphyllin, Egifilin, Elan, Elixophyllin, Elixophylline, Etheophyl, Euphylong, GM 501 SpermMobil, Insanovin, LaBID, Lanophyllin, Lasma, Liquophylline, NSC 2066, Nuelin, Optiphyllin, Parkophyllin, Physpan, Pro-Vent, Pseudotheophylline, PulmiDur, Pulmo-Timelets, Quibron T, Quibron T/SR, Respbid, Respicur, Slo-Bid, Slo-Phyllin, Solosin, Somophyllin CRT, Somophyllin T, Spophyllin retard, Sustaire, Talotren, Telb-DS, Telbans, Telbans Dry Syrup, Teocen 200, Teonova, Teotard, Teotard 350, Tesona, Theo 24, Theo-DS, Theo-Dur, Theo-Nite, Theo-Sav, Theobid, Theobid Duracap, Theochron, Theoclear, Theodel, Theodrip, Theodur Dry Syrup, Theofol, Theograd, Theolair, Theon, Theona P, Theopek, Theophyl, Theophyllin, Theoplus, Theostat, Theotard, Theovent, Throphylline, Unicontin CR, Unifyl, Unilong, Uniphyl, Uniphyllin, Xanthium, phylobid
IUPAC Name 1,3-dimethyl-7H-purine-2,6-dione
SMILES CN1C(=O)N(C)c2nc[nH]c2C1=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C7H8N4O2/c1-10-5-4(8-3-9-5)6(12)11(2)7(10)13/h3H,1-2H3,(H,8,9)
Formula Weight 180.0647
Percent Purity >95%
Chemical Name or Material Theophylline
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.