missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Theophylline, Mikromol™
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Description
Theophylline
Specifications
Specifications
| Chemical Name or Material | Theophylline |
| CAS | 58-55-9 |
| Analyte or Component Names | Theophylline |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H8 N4 O2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 1,3-Dimethyl-3,7-dihydro-1H-purine-2,6-dione, Theophylline, Diprophylline Imp. B (EP), Pentoxifylline Imp. C (EP), Dimenhydrinate Imp. A (EP), Caffeine Imp. A (EP), 1H-Purine-2,6-dione, 3,9-dihydro-1,3-dimethyl-, 1H-Purine-2,6-dione, 3,7-dihydro-1,3-dimethyl- (9CI), Theophylline (8CI), 1,3-Dimethyl-1H-purine-2,6(3H,7H)-dione, 1,3-Dimethylxanthine, 3,9-Dihydro-1,3-dimethyl-1H-purine-2,6-dione, Accurbron, Aerobin, Aerolate, Afonilm, Apnecut, Armophylline, Asmax, Austyn, Bilordyl, Brochoretard, Bronkodyl, Cetraphylline, Chronophyllin, Constant T, Controfilina 200, Diffumal, Duraphyl, Duraphyllin, Egifilin, Elan, Elixophyllin, Elixophylline, Etheophyl, Euphylong, GM 501 SpermMobil, Insanovin, LaBID, Lanophyllin, Lasma, Liquophylline, NSC 2066, Nuelin, Optiphyllin, Parkophyllin, Physpan, Pro-Vent, Pseudotheophylline, PulmiDur, Pulmo-Timelets, Quibron T, Quibron T/SR, Respbid, Respicur, Slo-Bid, Slo-Phyllin, Solosin, Somophyllin CRT, Somophyllin T, Spophyllin retard, Sustaire, Talotren, Telb-DS, Telbans, Telbans Dry Syrup, Teocen 200, Teonova, Teotard, Teotard 350, Tesona, Theo 24, Theo-DS, Theo-Dur, Theo-Nite, Theo-Sav, Theobid, Theobid Duracap, Theochron, Theoclear, Theodel, Theodrip, Theodur Dry Syrup, Theofol, Theograd, Theolair, Theon, Theona P, Theopek, Theophyl, Theophyllin, Theoplus, Theostat, Theotard, Theovent, Throphylline, Unicontin CR, Unifyl, Unilong, Uniphyl, Uniphyllin, Xanthium, phylobid, Caffeine Impurity A, Dimenhydrinate Impurity A, Diprophylline Impurity B, Pentoxifylline Impurity C |
| InChI Formula | InChI=1S/C7H8N4O2/c1-10-5-4(8-3-9-5)6(12)11(2)7(10)13/h3H,1-2H3,(H,8,9) |
| SMILES | CN1C(=O)N(C)c2nc[nH]c2C1=O |
| IUPAC Name | 1,3-dimethyl-7H-purine-2,6-dione |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction