missing translation for 'onlineSavingsMsg'
Learn More

Theophylline, LoGiCal

Product Code. 30457915
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457915 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457915 Supplier LoGiCal Supplier No. LGCFOR0176.00

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Theophylline
CAS 58-55-9
Analyte or Component Names Theophylline
Shipping Condition Room Temperature
Molecular Formula C7 H8 N4 O2
Certifications/Compliance ISO 9001
Synonym 1,3-Dimethyl-3,7-dihydro-1H-purine-2,6-dione, Theophylline, Diprophylline Imp. B (EP), Pentoxifylline Imp. C (EP), Dimenhydrinate Imp. A (EP), Caffeine Imp. A (EP), 1H-Purine-2,6-dione, 3,9-dihydro-1,3-dimethyl-, 1H-Purine-2,6-dione, 3,7-dihydro-1,3-dimethyl- (9CI), Theophylline (8CI), 1,3-Dimethyl-1H-purine-2,6(3H,7H)-dione, 1,3-Dimethylxanthine, 3,9-Dihydro-1,3-dimethyl-1H-purine-2,6-dione, Accurbron, Aerobin, Aerolate, Afonilm, Apnecut, Armophylline, Asmax, Austyn, Bilordyl, Brochoretard, Bronkodyl, Cetraphylline, Chronophyllin, Constant T, Controfilina 200, Diffumal, Duraphyl, Duraphyllin, Egifilin, Elan, Elixophyllin, Elixophylline, Etheophyl, Euphylong, GM 501 SpermMobil, Insanovin, LaBID, Lanophyllin, Lasma, Liquophylline, NSC 2066, Nuelin, Optiphyllin, Parkophyllin, Physpan, Pro-Vent, Pseudotheophylline, PulmiDur, Pulmo-Timelets, Quibron T, Quibron T/SR, Respbid, Respicur, Slo-Bid, Slo-Phyllin, Solosin, Somophyllin CRT, Somophyllin T, Spophyllin retard, Sustaire, Talotren, Telb-DS, Telbans, Telbans Dry Syrup, Teocen 200, Teonova, Teotard, Teotard 350, Tesona, Theo 24, Theo-DS, Theo-Dur, Theo-Nite, Theo-Sav, Theobid, Theobid Duracap, Theochron, Theoclear, Theodel, Theodrip, Theodur Dry Syrup, Theofol, Theograd, Theolair, Theon, Theona P, Theopek, Theophyl, Theophyllin, Theoplus, Theostat, Theotard, Theovent, Throphylline, Unicontin CR, Unifyl, Unilong, Uniphyl, Uniphyllin, Xanthium, phylobid, Caffeine Impurity A, Dimenhydrinate Impurity A, Diprophylline Impurity B, Pentoxifylline Impurity C
InChI Formula InChI=1S/C7H8N4O2/c1-10-5-4(8-3-9-5)6(12)11(2)7(10)13/h3H,1-2H3,(H,8,9)
Solution Type Neat
SMILES CN1C(=O)N(C)c2nc[nH]c2C1=O
IUPAC Name 1,3-dimethyl-7H-purine-2,6-dione
Industry Type Clinical Testing
Molecular Weight (g/mol) 180.16
Quantity 10 mg
Formula Weight 180.0647 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.