missing translation for 'onlineSavingsMsg'
Learn More

Theophylline EP Impurity B (Pentoxifylline EP Impurity B), Mikromol™

Product Code. 17960816
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17960816 100 mg 100mg
17911846 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17960816 Supplier Mikromol Supplier No. MM0176.03
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Theophylline EP Impurity B (Pentoxifylline EP Impurity B)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Methyl-3,7-dihydro-1H-purine-2,6-dione (3-Methylxanthine)
CAS 1076-22-8
Analyte or Component Names 3-Methyl-3,7-dihydro-1H-purine-2,6-dione (3-Methylxanthine)
Shipping Condition Room Temperature
Molecular Formula C6 H6 N4 O2
Certifications/Compliance ISO 17025
Synonym Pentoxifylline Imp. B (EP), Theophylline Imp. B (EP), Theophylline-Ethylenediamine Anhydrous Imp. B (EP), Theophylline USP Related Compound B, Theophylline USP RC B, 3-Methyl-3,7-dihydro-1H-purine-2,6-dione, 3-Methylxanthine, Theophylline-Ethylenediamine Imp. B (EP), Theophylline Related Compound B, Pentoxifylline Impurity B, Theophylline Impurity B, Theophylline-Ethylenediamine Impurity B
InChI Formula InChI=1S/C6H6N4O2/c1-10-4-3(7-2-8-4)5(11)9-6(10)12/h2H,1H3,(H,7,8)(H,9,11,12)
SMILES CN1C(=O)NC(=O)c2[nH]cnc12
IUPAC Name 3-methyl-7H-purine-2,6-dione
Molecular Weight (g/mol) 166.14
Quantity 100 mg
Formula Weight 166.0491 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.