missing translation for 'onlineSavingsMsg'
Learn More

Theophylline-7-acetic Acid, TRC

Product Code. 30138116
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138116 10 g 10g
30136214 25 g 25g
30008562 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138116 Supplier Toronto Research Chemicals Supplier No. TRCT34386010G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 652-37-9
Molecular Formula C9 H10 N4 O4
Molecular Weight (g/mol) 238.2
Synonym 7H-Purine-7-acetic acid, 1,2,3,6-tetrahydro-1,3-dimethyl-2,6-dioxo-Purine-7-acetic acid, 1,2,3,6-tetrahydro-1,3-dimethyl-2,6-dioxo- (6CI,7CI,8CI), 1,2,3,6-Tetrahydro-1,3-dimethyl-2,6-dioxopurine-7-acetic acid, 7-(Carboxymethyl)theophylline, 7-Theophyllineacetic acid, 7-Theophyllinylacetic acid, Acefylline, Acephylline, Carboxymethyltheophylline, N-7-Theophyllineacetic acid, NSC 52996, Theophyllineacetic acid
IUPAC Name 2-(1,3-dimethyl-2,6-dioxopurin-7-yl)acetic acid
SMILES CN1C(=O)N(C)c2ncn(CC(=O)O)c2C1=O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C9H10N4O4/c1-11-7-6(8(16)12(2)9(11)17)13(4-10-7)3-5(14)15/h4H,3H2,1-2H3,(H,14,15)
Formula Weight 238.0702
Chemical Name or Material Acefylline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.